Learn More
Medchemexpress LLC 3-indoxyl butyrate | 4346-15-0 | MFCD00022718 | 99.9% | 203.24 | C12H13NO2 | 250 MG
SDP
Catalog No. 5000281843
Encompass_Preferred
3-indoxyl butyrate is a chromogenic biochemical assay reagent used as a substrate for carboxylesterase (CES). Upon enzymatic cleavage it produces a blue precipitate, enabling visual or colorimetric detection of CES activity. Key identifiers include CAS 4346-15-0, formula C12H13NO2, and molecular weight 203.24 g·mol⁻1.
  • Chromogenic substrate for carboxylesterase that produces a blue precipitate upon enzymatic cleavage.
  • High purity (≈99.9%) suitable for biochemical assays and analytical work.
  • Small-molecule organic compound; CAS 4346-15-0; formula C12H13NO2; molecular weight 203.24 g·mol⁻1.
  • Supplied as a solid for preparation of stock solutions in assay workflows.
Catalog No. 50-002-81843 Supplier Medchemexpress LLC Supplier No. HYW341421250MG
3-indoxyl butyrate is a chromogenic biochemical assay reagent used as a substrate for carboxylesterase (CES). Upon enzymatic cleavage it produces a blue precipitate, enabling visual or colorimetric detection of CES activity. Key identifiers include CAS 4346-15-0, formula C12H13NO2, and molecular weight 203.24 g·mol⁻1.
  • Chromogenic substrate for carboxylesterase that produces a blue precipitate upon enzymatic cleavage.
  • High purity (≈99.9%) suitable for biochemical assays and analytical work.
  • Small-molecule organic compound; CAS 4346-15-0; formula C12H13NO2; molecular weight 203.24 g·mol⁻1.
  • Supplied as a solid for preparation of stock solutions in assay workflows.