Learn More
Medchemexpress LLC 3-indoxyl butyrate | 4346-15-0 | MFCD00022718 | 99.9% | 203.24 g·mol⁻1 | C12H13NO2 | 500 MG
SDP
Catalog No. 5000282543
Encompass_Preferred
3-Indoxyl butyrate is a chromogenic substrate for carboxylesterase (CES). Upon enzymatic cleavage it produces an insoluble blue precipitate, enabling colorimetric and histochemical detection of esterase activity in biochemical and tissue assays. Supplied as a white to off-white solid with high listed purity, it is intended for research use only.
  • Chromogenic detection of carboxylesterase activity.
  • Produces an insoluble blue precipitate upon enzymatic cleavage.
  • High purity suitable for analytical and biochemical assays.
  • Solid form for easy handling and accurate weighing.
  • Stable when stored protected from light at recommended temperatures.
Catalog No. 50-002-82543 Supplier Medchemexpress LLC Supplier No. HYW341421500MG
3-Indoxyl butyrate is a chromogenic substrate for carboxylesterase (CES). Upon enzymatic cleavage it produces an insoluble blue precipitate, enabling colorimetric and histochemical detection of esterase activity in biochemical and tissue assays. Supplied as a white to off-white solid with high listed purity, it is intended for research use only.
  • Chromogenic detection of carboxylesterase activity.
  • Produces an insoluble blue precipitate upon enzymatic cleavage.
  • High purity suitable for analytical and biochemical assays.
  • Solid form for easy handling and accurate weighing.
  • Stable when stored protected from light at recommended temperatures.