Learn More
Medchemexpress LLC 3-indoxyl butyrate | 4346-15-0 | MFCD00022718 | 99.9% | 203.24 g/mol | C12H13NO2 | 1 G
SDP
Catalog No. 5000282347
Encompass_Preferred
3-Indoxyl butyrate is a chromogenic substrate for carboxylesterase (CES) used in biochemical and enzymatic assays; enzymatic cleavage produces a blue precipitate that enables visual or colorimetric detection. The compound is supplied at high purity for assay development and detection workflows.
  • Chromogenic substrate for carboxylesterase producing a blue precipitate upon cleavage.
  • High purity: 99.9%.
  • Suitable for visual and colorimetric assay readouts.
  • Stable when stored at -20°C protected from light.
  • Available in multiple packaging sizes for research use.
Catalog No. 50-002-82347 Supplier Medchemexpress LLC Supplier No. HYW3414211G
3-Indoxyl butyrate is a chromogenic substrate for carboxylesterase (CES) used in biochemical and enzymatic assays; enzymatic cleavage produces a blue precipitate that enables visual or colorimetric detection. The compound is supplied at high purity for assay development and detection workflows.
  • Chromogenic substrate for carboxylesterase producing a blue precipitate upon cleavage.
  • High purity: 99.9%.
  • Suitable for visual and colorimetric assay readouts.
  • Stable when stored at -20°C protected from light.
  • Available in multiple packaging sizes for research use.