Learn More
Medchemexpress LLC 3-indoxyl butyrate | 4346-15-0 | MFCD00022718 | 99.9% | 203.24 g/mol | C12H13NO2 | 100 MG
SDP
Catalog No. 5000280934
Encompass_Preferred
3-Indoxyl butyrate is a chromogenic substrate for carboxylesterase (CES) that produces an insoluble blue precipitate upon enzymatic cleavage, enabling visualization and quantification of esterase activity in biochemical and histochemical assays.
  • Produces a blue precipitate upon cleavage for clear visual detection.
  • Suitable for colorimetric and histochemical assays.
  • High purity for reproducible results.
  • Compatible with enzymatic detection workflows and staining protocols.
  • Available in multiple pack sizes for research scalability.
Catalog No. 50-002-80934 Supplier Medchemexpress LLC Supplier No. HYW341421100MG
3-Indoxyl butyrate is a chromogenic substrate for carboxylesterase (CES) that produces an insoluble blue precipitate upon enzymatic cleavage, enabling visualization and quantification of esterase activity in biochemical and histochemical assays.
  • Produces a blue precipitate upon cleavage for clear visual detection.
  • Suitable for colorimetric and histochemical assays.
  • High purity for reproducible results.
  • Compatible with enzymatic detection workflows and staining protocols.
  • Available in multiple pack sizes for research scalability.