Learn More
Medchemexpress LLC 3-indoxyl butyrate | 4346-15-0 | MFCD00022718 | 99.9% | 203.24 g/mol | C12H13NO2 | 50 MG
SDP
Catalog No. 5000282345
Encompass_Preferred
3-Indoxyl butyrate is a chromogenic biochemical reagent used to detect carboxylesterase (CES) activity. Upon enzymatic cleavage it produces a blue precipitate, enabling colorimetric or histochemical visualization of CES in biochemical assays. The compound is supplied at high purity for research applications and is intended for analytical and detection workflows.
Catalog No. 50-002-82345 Supplier Medchemexpress LLC Supplier No. HYW34142150MG
3-Indoxyl butyrate is a chromogenic biochemical reagent used to detect carboxylesterase (CES) activity. Upon enzymatic cleavage it produces a blue precipitate, enabling colorimetric or histochemical visualization of CES in biochemical assays. The compound is supplied at high purity for research applications and is intended for analytical and detection workflows.