Learn More
Alkali Scientific 3 Indoxyl β D Glucoside [IBDG] [3 Indoxyl β D Glucopyranoside], 1 G
SDP
Catalog No. 5000659558
Encompass_Preferred
3-Indoxyl β-D-Glucoside (IBDG) is a chromogenic substrate used for the detection of β-glucosidase activity in biochemical assays. When β-glucosidase hydrolyzes IBDG, an indigo-colored product is produced, which serves as a visible marker for enzyme activity. The 1g format offers a larger amount of reagent suitable for high-throughput applications or larger-scale experiments. IBDG is commonly used in molecular biology, microbiology, and enzyme research for monitoring gene expression, screening for β-glucosidase activity, and studying the regulation of metabolic pathways involving glucosidases. The colorimetric result makes it ideal for a variety of assays requiring visual confirmation of enzyme activity.
Catalog No. 50-006-59558 Supplier Alkali Scientific Supplier No. IBD003CP
3-Indoxyl β-D-Glucoside (IBDG) is a chromogenic substrate used for the detection of β-glucosidase activity in biochemical assays. When β-glucosidase hydrolyzes IBDG, an indigo-colored product is produced, which serves as a visible marker for enzyme activity. The 1g format offers a larger amount of reagent suitable for high-throughput applications or larger-scale experiments. IBDG is commonly used in molecular biology, microbiology, and enzyme research for monitoring gene expression, screening for β-glucosidase activity, and studying the regulation of metabolic pathways involving glucosidases. The colorimetric result makes it ideal for a variety of assays requiring visual confirmation of enzyme activity.