Learn More
Alkali Scientific 3 Indoxyl β D Glucoside [IBDG] [3 Indoxyl β D Glucopyranoside], 10 G
SDP
Catalog No. 5000659598
Encompass_Preferred
3-Indoxyl β-D-Glucoside (IBDG) is a highly sensitive chromogenic substrate used for the detection of β-glucosidase activity in biochemical assays. The hydrolysis of IBDG by β-glucosidase results in the production of an indigo-colored compound, providing a visual indicator of enzyme activity. The 10g quantity is ideal for large-scale experiments or long-term studies. IBDG is commonly used in molecular biology, microbiology, and biochemical research to detect and quantify β-glucosidase activity in various organisms and samples. This reagent offers a convenient, easy-to-interpret method for screening enzyme activity, studying gene expression systems, and analyzing enzymatic reactions in a wide range of applications.
Catalog No. 50-006-59598 Supplier Alkali Scientific Supplier No. IBD004CP
3-Indoxyl β-D-Glucoside (IBDG) is a highly sensitive chromogenic substrate used for the detection of β-glucosidase activity in biochemical assays. The hydrolysis of IBDG by β-glucosidase results in the production of an indigo-colored compound, providing a visual indicator of enzyme activity. The 10g quantity is ideal for large-scale experiments or long-term studies. IBDG is commonly used in molecular biology, microbiology, and biochemical research to detect and quantify β-glucosidase activity in various organisms and samples. This reagent offers a convenient, easy-to-interpret method for screening enzyme activity, studying gene expression systems, and analyzing enzymatic reactions in a wide range of applications.