Learn More
Alkali Scientific 3 Indoxyl β D Glucoside [IBDG] [3 Indoxyl β D Glucopyranoside], 500 Mg
SDP
Catalog No. 5000659557
Encompass_Preferred
3-Indoxyl β-D-Glucoside (IBDG) is a sensitive chromogenic substrate used for detecting β-glucosidase activity in biochemical assays. This reagent produces a distinctive indigo color when hydrolyzed by β-glucosidase, providing a visual indicator of enzyme activity. The 500mg quantity is ideal for larger experiments, high-throughput screening, or long-term studies. IBDG is widely used in microbiology and molecular biology for monitoring enzyme activity, conducting reporter assays, and studying gene expression systems. It offers an easy-to-use, reliable method for detecting β-glucosidase activity in cultured cells, bacterial colonies, and other biological samples.
Catalog No. 50-006-59557 Supplier Alkali Scientific Supplier No. IBD002CP
3-Indoxyl β-D-Glucoside (IBDG) is a sensitive chromogenic substrate used for detecting β-glucosidase activity in biochemical assays. This reagent produces a distinctive indigo color when hydrolyzed by β-glucosidase, providing a visual indicator of enzyme activity. The 500mg quantity is ideal for larger experiments, high-throughput screening, or long-term studies. IBDG is widely used in microbiology and molecular biology for monitoring enzyme activity, conducting reporter assays, and studying gene expression systems. It offers an easy-to-use, reliable method for detecting β-glucosidase activity in cultured cells, bacterial colonies, and other biological samples.