Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Indoxyl Phosphate, Disodium Salt, >98%, MP Biomedicals™

Catalog No. ICN15416783 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15416783 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15416783 Supplier MP Biomedicals, Inc Supplier No. 0215416783
Add to Cart
Add to Cart

3-Indoxyl Phosphate Disodium Salt is used as a histochemical substrate to demonstrate alkaline phosphatase

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3318-43-2
Molecular Formula C8H6NNa2O4P
Molecular Weight (g/mol) 257.092
MDL Number MFCD00040646
InChI Key WCSKWBKPKXJWEG-UHFFFAOYSA-L
Synonym 3-indoxyl phosphate disodium salt, disodium indol-3-yl phosphate, indol-3-yl sodium phosphate, disodium 1h-indol-3-yl phosphate, unii-p1w2d6403e, indoxylphosphate, disodium salt, sodium 1h-indol-3-yl phosphate, 1h-indol-3-ol, dihydrogen phosphate ester , disodium salt, ip, na2, 3-indoxyl phosphate
PubChem CID 76830
IUPAC Name disodium;1H-indol-3-yl phosphate
SMILES C1=CC=C2C(=C1)C(=CN2)OP(=O)([O-])[O-].[Na+].[Na+]

Specifications

Color Gray
Assay Percent Range > 98%
Quantity 250 mg
Formula Weight 257.1
Physical Form Powder
Chemical Name or Material 3-Indoxyl Phosphate, Disodium Salt, FTIR
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.