Learn More
Cayman Chemical 3-Indoxyl Phosphate (sodium salt), Purity: ≥95%, CAS: 3318-43-2, 1 g
SDP
Catalog No. 502947670
Encompass_Preferred
A chromogenic and electrochemical substrate for alkaline phosphatase; enzymatic cleavage by alkaline phosphatase releases 3-indoxyl and is subsequently oxidized to produce insoluble indigo, which can be quantified by colorimetric detection at 660 nm; can be converted to water-soluble leucoindigo by reduction of indigo in alkaline solution by sodium dithionite, which can be quantified by colorimetric detection at 415 nm or voltametric detection; has been used in ELISAs
Catalog No. 50-294-7670 Supplier Cayman Chemical Supplier No. 339361 G
A chromogenic and electrochemical substrate for alkaline phosphatase; enzymatic cleavage by alkaline phosphatase releases 3-indoxyl and is subsequently oxidized to produce insoluble indigo, which can be quantified by colorimetric detection at 660 nm; can be converted to water-soluble leucoindigo by reduction of indigo in alkaline solution by sodium dithionite, which can be quantified by colorimetric detection at 415 nm or voltametric detection; has been used in ELISAs