Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Iodo-1-tetrahydropyran-2-yl-pyrazole-4-carbaldehyde | 1627924-19-9 | 99.8% | 306.10 | 5 G
SDP

Catalog No. 5000403692
Encompass_Preferred

3-Iodo-1-tetrahydropyran-2-yl-pyrazole-4-carbaldehyde is a drug intermediate for the synthesis of various active compounds. It is sold for research purposes only and not for patient use.

  • Solid appearance
  • Off-white to light yellow color
  • Store at -20°C, protect from light, and under nitrogen
  • Soluble in DMSO: 100 mg/mL
  • For research purposes only

Catalog No. 50-004-03692 Supplier Medchemexpress LLC Supplier No. HY415145G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Iodo-1-tetrahydropyran-2-yl-pyrazole-4-carbaldehyde is a drug intermediate for the synthesis of various active compounds. It is sold for research purposes only and not for patient use.

  • Solid appearance
  • Off-white to light yellow color
  • Store at -20°C, protect from light, and under nitrogen
  • Soluble in DMSO: 100 mg/mL
  • For research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.