Learn More
Medchemexpress LLC 3-Iodo-1-tetrahydropyran-2-yl-pyrazole-4-carbaldehyde | 1627924-19-9 | >99.8% | 306.10 | 500 MG
SDP
Catalog No. 5000404898
Encompass_Preferred
3-Iodo-1-tetrahydropyran-2-yl-pyrazole-4-carbaldehyde is identified as a drug intermediate utilized in the synthesis of various active compounds. It appears as an off-white to light yellow solid. The compound demonstrates high solubility in DMSO, with specific guidelines for preparing stock solutions and handling to maintain product integrity.
  • Used in the synthesis of active compounds.
  • Appears as an off-white to light yellow solid.
  • Soluble in DMSO.
Catalog No. 50-004-04898 Supplier Medchemexpress LLC Supplier No. HY41514500MG
3-Iodo-1-tetrahydropyran-2-yl-pyrazole-4-carbaldehyde is identified as a drug intermediate utilized in the synthesis of various active compounds. It appears as an off-white to light yellow solid. The compound demonstrates high solubility in DMSO, with specific guidelines for preparing stock solutions and handling to maintain product integrity.
  • Used in the synthesis of active compounds.
  • Appears as an off-white to light yellow solid.
  • Soluble in DMSO.