Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodo-4-methylbenzoic acid, 97%, Thermo Scientific™

Catalog No. AC249790250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC249790250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC249790250 Supplier Thermo Scientific Chemicals Supplier No. 249790250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82998-57-0
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.046
InChI Key LDDHMKANNXWUAK-UHFFFAOYSA-N
Synonym 3-iodo-4-methyl-benzoic acid, benzoic acid, 3-iodo-4-methyl, zlchem 953, 3-iodo-p-toluicacid, pubchem3947, 4-carboxy-2-iodotoluene, akos bbb/227, ksc449e9t, 3-iodo-4-methyl benzoic acid, paragos 440021
PubChem CID 621640
IUPAC Name 3-iodo-4-methylbenzoic acid
SMILES CC1=C(C=C(C=C1)C(=O)O)I

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 207°C to 212°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula IC6H3(CH3)CO2H
Quantity 25 g
Formula Weight 262.04
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodo-4-methylbenzoic acid, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.