Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodo-4-nitroanisole, 97%

Catalog No. AC326320250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC326320250 25 g Glass bottle
AC326320050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC326320250 Supplier Thermo Scientific Chemicals Supplier No. 326320250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 214279-40-0 | C7H6INO3 | 279.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 214279-40-0
Molecular Formula C7H6INO3
Molecular Weight (g/mol) 279.03
MDL Number MFCD01320682
InChI Key NLTAHTYBZZSCIT-UHFFFAOYSA-N
Synonym 3-iodo-4-nitroanisole, 4-nitro-3-iodoanisole, benzene, 2-iodo-4-methoxy-1-nitro, 1-nitro-2-iodo-4-methoxybenzene, 3-iodo-4-nitrophenyl methyl ether, benzene,2-iodo-4-methoxy-1-nitro
PubChem CID 2733456
IUPAC Name 2-iodo-4-methoxy-1-nitrobenzene
SMILES COC1=CC=C(C(I)=C1)[N+]([O-])=O

Specifications

Melting Point 68.0°C to 73.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 279.03
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3-Iodo-4-nitroanisole
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.