Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-iodo-L-phenylalanine | 20846-39-3 | MFCD01860664 | >97.0% | 291.09 g/mol | C9H10INO2 | 1 G
SDP

Catalog No. 5000280626
Encompass_Preferred

3-Iodo-L-phenylalanine is an iodinated, noncanonical derivative of L-phenylalanine used in biochemical research, genetic code expansion, and protein engineering. Supplied as a solid for laboratory use, it is used in peptide synthesis and studies of protein translation and cell proliferation.

  • Iodinated noncanonical amino acid
  • Useful for genetic code expansion and codon reassignment studies
  • Suitable for protein engineering and peptide synthesis
  • White to off-white solid with high purity
  • Purity >97.0%
  • Molecular formula C9H10INO2 and molecular weight 291.09 g/mol
  • Supplied as 1 G package for laboratory use

Catalog No. 50-002-80626 Supplier Medchemexpress LLC Supplier No. HYW2597671G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Iodo-L-phenylalanine is an iodinated, noncanonical derivative of L-phenylalanine used in biochemical research, genetic code expansion, and protein engineering. Supplied as a solid for laboratory use, it is used in peptide synthesis and studies of protein translation and cell proliferation.

  • Iodinated noncanonical amino acid
  • Useful for genetic code expansion and codon reassignment studies
  • Suitable for protein engineering and peptide synthesis
  • White to off-white solid with high purity
  • Purity >97.0%
  • Molecular formula C9H10INO2 and molecular weight 291.09 g/mol
  • Supplied as 1 G package for laboratory use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.