Learn More
Medchemexpress LLC 3-iodo-L-phenylalanine | 20846-39-3 | MFCD01860664 | 99.3% | 291.09 g·mol⁻1 | C9H10INO2 | 250 MG
SDP
Catalog No. 5000281388
Encompass_Preferred
3-Iodo-L-phenylalanine is a noncanonical amino acid bearing an iodine substituent at the 3-position of the phenyl ring. It is supplied as a high-purity research reagent intended for peptide synthesis, genetic code expansion, and other biochemical studies that require halogenated phenylalanine analogs.
  • High purity suitable for research applications.
  • Iodine substitution at the 3-position enables site-specific modification studies.
  • Compatible with peptide synthesis and genetic code expansion workflows.
  • Available in laboratory-scale quantities for experimental use.
  • Documentation includes CAS, molecular formula, and molecular weight.
Catalog No. 50-002-81388 Supplier Medchemexpress LLC Supplier No. HYW259767250MG
3-Iodo-L-phenylalanine is a noncanonical amino acid bearing an iodine substituent at the 3-position of the phenyl ring. It is supplied as a high-purity research reagent intended for peptide synthesis, genetic code expansion, and other biochemical studies that require halogenated phenylalanine analogs.
  • High purity suitable for research applications.
  • Iodine substitution at the 3-position enables site-specific modification studies.
  • Compatible with peptide synthesis and genetic code expansion workflows.
  • Available in laboratory-scale quantities for experimental use.
  • Documentation includes CAS, molecular formula, and molecular weight.