Learn More
Medchemexpress LLC 3-iodo-L-phenylalanine | 20846-39-3 | MFCD01860664 | 99.3% | 291.09 g·mol⁻¹ | C9H10INO2 | 500 MG
SDP
Catalog No. 5000281671
Encompass_Preferred
3-Iodo-L-phenylalanine is a noncanonical amino acid used in genetic code expansion and protein engineering to incorporate an iodinated phenylalanine residue into peptides and proteins for structural, functional, or labeling studies. It is supplied as a solid with high stated purity and is intended for research use only.
  • High purity (~99.3%) suitable for analytical and preparative work.
  • Molecular weight 291.09 g·mol⁻¹ for accurate formulation and calculations.
  • Solid, white to off-white appearance for easy handling and visual inspection.
  • Recommended cold storage and protection from light to preserve stability.
  • Available in multiple package sizes, including 500 mg, for flexible experimental needs.
Catalog No. 50-002-81671 Supplier Medchemexpress LLC Supplier No. HYW259767500MG
3-Iodo-L-phenylalanine is a noncanonical amino acid used in genetic code expansion and protein engineering to incorporate an iodinated phenylalanine residue into peptides and proteins for structural, functional, or labeling studies. It is supplied as a solid with high stated purity and is intended for research use only.
  • High purity (~99.3%) suitable for analytical and preparative work.
  • Molecular weight 291.09 g·mol⁻¹ for accurate formulation and calculations.
  • Solid, white to off-white appearance for easy handling and visual inspection.
  • Recommended cold storage and protection from light to preserve stability.
  • Available in multiple package sizes, including 500 mg, for flexible experimental needs.