Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodobenzaldehyde, 98%

Catalog No. AAL1744703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1744703 1 g
AAL1744706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1744703 Supplier Thermo Scientific Chemicals Supplier No. L1744703
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 696-41-3 | C7H5IO | 232.02 g/mol

3-Iodobenzaldehyde is used as an organic reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodobenzaldehyde is used as an organic reagent.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. Store under inert gas. It is sensitive to air and light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 696-41-3
Molecular Formula C7H5IO
Molecular Weight (g/mol) 232.02
MDL Number MFCD00039573
InChI Key RZODAQZAFOBFLS-UHFFFAOYSA-N
Synonym 3-iodo-benzaldehyde, 3-iodbenzaldehyd, benzaldehyde, 3-iodo, 3-iodo benzaldehyde, 3-iodanylbenzaldehyde, pubchem3917, acmc-1atsd, 1-formyl-3-iodobenzene, m-iodobenzaldehyde, 3-iodobenzaldehyde
PubChem CID 252610
IUPAC Name 3-iodobenzaldehyde
SMILES C1=CC(=CC(=C1)I)C=O

Specifications

Melting Point 55°C to 57°C
Boiling Point 124°C to 125°C (13 mmHg)
Quantity 1 g
Beilstein 1854653
Sensitivity Air and light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 232.02
Percent Purity 98%
Chemical Name or Material 3-Iodobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.