Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (3-Iodopropyl)Trimethoxysilane

Catalog No. 5803525MLF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-035-25MLF 25 mL
58-035-100MLF 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 58-035-25MLF Supplier Sigma Aldrich Supplier No. 5803525MLF
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(3-iodopropyl)trimethoxysilane may be used to organically modify mesoporous silica support by grafting method.{85} The product may be used to synthesize heavy atom-concentratedorganically modified silicananoparticles.{86}

Specifications

CAS 14867-28-8
Density 1.482 g/mL at 20°C
Molecular Formula C6H15IO3Si
Refractive Index n20/D 1.473
Linear Formula C6H15IO3Si
MDL Number MFCD00054838
Quantity 25 mL
Molecular Weight (g/mol) 290.17 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.