Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodopyridine, 99%, Thermo Scientific™

Catalog No. AC137440050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC137440050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC137440050 Supplier Thermo Scientific Chemicals Supplier No. 137440050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-90-7
Molecular Formula C5H4IN
Molecular Weight (g/mol) 204.998
InChI Key XDELKSRGBLWMBA-UHFFFAOYSA-N
Synonym 3-iodo pyridine, 3-iodo-pyridine, pyridine, 3-iodo, 3-iodpyridin, 3-iodopyridine, pubchem6634, acmc-1bvap, ksc490s0t
PubChem CID 70714
IUPAC Name 3-iodopyridine
SMILES C1=CC(=CN=C1)I

Specifications

Melting Point 53.0°C to 56.0°C
Flash Point 106°C
Quantity 5 g
Formula Weight 205
Chemical Name or Material 3-Iodopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.