Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodothiophene, 97%

Catalog No. AC443380010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC443380010 1 g
AC443380050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC443380010 Supplier Thermo Scientific Chemicals Supplier No. 443380010
Add to Cart

CAS: 10486-61-0 | C4H3IS | 210.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10486-61-0
Molecular Formula C4H3IS
Molecular Weight (g/mol) 210.03
MDL Number MFCD00037901
InChI Key WGKRMQIQXMJVFZ-UHFFFAOYSA-N
Synonym thiophene, 3-iodo, 3-iodothiophene, stab. with copper, 3-lodothiophene, 3-iodo-thiophene, 3-iodanylthiophene, 3-thienyl iodide, thien-3-yl iodide, pubchem16213, acmc-2098dn
PubChem CID 66332
IUPAC Name 3-iodothiophene
SMILES IC1=CSC=C1

Specifications

Melting Point -13.0°C
Density 2.0660g/mL
Boiling Point 195.0°C
Flash Point 78°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 2.066
Formula Weight 210.03
Percent Purity 97%
Chemical Name or Material 3-Iodothiophene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw
EINECSNumber 234-009-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.