Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-isobutyl-1-methylxanthine | 28822-58-4 | MFCD00005584 | 100.0% | 222.24 | C10H14N4O2 | 25MG
SDP

Catalog No. 5000235617
Encompass_Preferred

IBMX (3-Isobutyl-1-methylxanthine) is a broad-spectrum phosphodiesterase (PDE) inhibitor used as a research reagent to elevate intracellular cyclic nucleotides by inhibiting multiple PDE isoforms. It is supplied as a stable solid with documented physicochemical, purity, and solubility data for preparation of in vitro and in vivo formulations.

  • Broad-spectrum PDE inhibitor with reported IC50s of 6.5 μM (PDE3), 26.3 μM (PDE4), and 31.7 μM (PDE5).
  • Molecular formula C10H14N4O2; molecular weight 222.24 g/mol.
  • High purity suitable for research (reported purities ~99.97-99.99%).
  • Soluble in DMSO (125 mg/mL) and compatible with published in vivo cosolvent formulations.
  • Available in multiple solid pack sizes, including 25 MG, for assay preparation.
  • Recommended storage: powder -20°C (short term refrigerated at 4°C), in solvent -80°C for long term.

Catalog No. 50-002-35617 Supplier Medchemexpress LLC Supplier No. HY1231825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IBMX (3-Isobutyl-1-methylxanthine) is a broad-spectrum phosphodiesterase (PDE) inhibitor used as a research reagent to elevate intracellular cyclic nucleotides by inhibiting multiple PDE isoforms. It is supplied as a stable solid with documented physicochemical, purity, and solubility data for preparation of in vitro and in vivo formulations.

  • Broad-spectrum PDE inhibitor with reported IC50s of 6.5 μM (PDE3), 26.3 μM (PDE4), and 31.7 μM (PDE5).
  • Molecular formula C10H14N4O2; molecular weight 222.24 g/mol.
  • High purity suitable for research (reported purities ~99.97-99.99%).
  • Soluble in DMSO (125 mg/mL) and compatible with published in vivo cosolvent formulations.
  • Available in multiple solid pack sizes, including 25 MG, for assay preparation.
  • Recommended storage: powder -20°C (short term refrigerated at 4°C), in solvent -80°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.