Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-isobutyl-1-methylxanthine | 28822-58-4 | MFCD00005584 | 222.24 g/mol | C10H14N4O2 | 10MG
SDP

Catalog No. 5000239005
Encompass_Preferred

IBMX (3-isobutyl-1-methylxanthine) is a GMP-grade broad-spectrum phosphodiesterase (PDE) inhibitor supplied in a 10 mg package for small-scale laboratory and process development use. It is intended as an auxiliary reagent in cell therapy manufacturing and related research applications, and exhibits inhibitory activity against multiple PDE isoforms.

  • GMP-grade material suitable for cell therapy workflows.
  • Broad-spectrum phosphodiesterase inhibitor active against multiple PDE isoforms.
  • Small 10 mg package for small-scale experiments and formulation development.
  • Chemical formula C10H14N4O2 and molecular weight 222.24 g/mol.
  • Certificate of analysis available for identity and purity documentation.

Catalog No. 50-002-39005 Supplier Medchemexpress LLC Supplier No. HY12318G10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IBMX (3-isobutyl-1-methylxanthine) is a GMP-grade broad-spectrum phosphodiesterase (PDE) inhibitor supplied in a 10 mg package for small-scale laboratory and process development use. It is intended as an auxiliary reagent in cell therapy manufacturing and related research applications, and exhibits inhibitory activity against multiple PDE isoforms.

  • GMP-grade material suitable for cell therapy workflows.
  • Broad-spectrum phosphodiesterase inhibitor active against multiple PDE isoforms.
  • Small 10 mg package for small-scale experiments and formulation development.
  • Chemical formula C10H14N4O2 and molecular weight 222.24 g/mol.
  • Certificate of analysis available for identity and purity documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.