Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-isobutyl-1-methylxanthine | 28822-58-4 | MFCD00005584 | >98.0% | 222.24 g/mol | C10H14N4O2 | 1G
SDP

Catalog No. 5000239004
Encompass_Preferred

IBMX (3-isobutyl-1-methylxanthine) is a non-selective cyclic nucleotide phosphodiesterase inhibitor used in research to raise intracellular cAMP and cGMP and to probe PKA/PKG signaling pathways. Supplied as a solid research reagent, it is used in biochemical assays and cell-based studies to pharmacologically increase cyclic nucleotide levels.

  • Non-selective phosphodiesterase inhibitor that elevates cAMP and cGMP.
  • Commonly used in cell signaling and biochemical assay workflows.
  • Purity >98.0% by HPLC (typical specification).
  • Chemical formula C10H14N4O2; molecular weight 222.24 g/mol.
  • CAS number 28822-58-4 for unambiguous identification.
  • Available in small-scale research pack sizes as a solid powder.

Catalog No. 50-002-39004 Supplier Medchemexpress LLC Supplier No. HY123181G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IBMX (3-isobutyl-1-methylxanthine) is a non-selective cyclic nucleotide phosphodiesterase inhibitor used in research to raise intracellular cAMP and cGMP and to probe PKA/PKG signaling pathways. Supplied as a solid research reagent, it is used in biochemical assays and cell-based studies to pharmacologically increase cyclic nucleotide levels.

  • Non-selective phosphodiesterase inhibitor that elevates cAMP and cGMP.
  • Commonly used in cell signaling and biochemical assay workflows.
  • Purity >98.0% by HPLC (typical specification).
  • Chemical formula C10H14N4O2; molecular weight 222.24 g/mol.
  • CAS number 28822-58-4 for unambiguous identification.
  • Available in small-scale research pack sizes as a solid powder.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.