Learn More
Medchemexpress LLC 3-isobutyl-1-methylxanthine | 28822-58-4 | MFCD00005584 | 100.0% | 222.24 g/mol | C10H14N4O2 | 500MG
SDP
Catalog No. 5000239142
Encompass_Preferred
IBMX (3-isobutyl-1-methylxanthine) is a non-selective phosphodiesterase (PDE) inhibitor supplied as an analytical standard for research use. It increases intracellular cAMP and cGMP by inhibiting multiple PDE isoforms and is provided as a high-purity solid for laboratory applications. Reported IC50 values include 6.5 μM (PDE3), 26.3 μM (PDE4), and 31.7 μM (PDE5).
  • Non-selective phosphodiesterase inhibitor.
  • Modulates intracellular cAMP and cGMP levels.
  • Useful for biochemical assays and cell-based studies.
  • Provided as a high-purity analytical standard for research.
  • Reported IC50s: PDE3 6.5 μM, PDE4 26.3 μM, PDE5 31.7 μM.
Catalog No. 50-002-39142 Supplier Medchemexpress LLC Supplier No. HY12318500MG
IBMX (3-isobutyl-1-methylxanthine) is a non-selective phosphodiesterase (PDE) inhibitor supplied as an analytical standard for research use. It increases intracellular cAMP and cGMP by inhibiting multiple PDE isoforms and is provided as a high-purity solid for laboratory applications. Reported IC50 values include 6.5 μM (PDE3), 26.3 μM (PDE4), and 31.7 μM (PDE5).
  • Non-selective phosphodiesterase inhibitor.
  • Modulates intracellular cAMP and cGMP levels.
  • Useful for biochemical assays and cell-based studies.
  • Provided as a high-purity analytical standard for research.
  • Reported IC50s: PDE3 6.5 μM, PDE4 26.3 μM, PDE5 31.7 μM.