Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-isobutyl-1-methylxanthine | 28822-58-4 | MFCD00005584 | 100.0% | 222.24 g/mol | C10H14N4O2 | 5G
SDP

Catalog No. 5000235616
Encompass_Preferred

IBMX (3-isobutyl-1-methylxanthine) is a broad-spectrum phosphodiesterase (PDE) inhibitor used in research to raise intracellular cyclic AMP and probe signaling pathways. It is provided as a solid reagent for biochemical and cell-based assays.

  • Broad-spectrum phosphodiesterase inhibitor.
  • Inhibitory activity: PDE3 IC50 6.5 μM, PDE4 IC50 26.3 μM, PDE5 IC50 31.7 μM.
  • Molecular formula C10H14N4O2; molecular weight 222.24 g/mol.
  • Supplied as a powdered reagent suitable for research use.
  • Storage: powder -20°C for long-term storage, 4°C for shorter storage.
  • Reported purity approximately 99.97% by HPLC.
  • Used to elevate intracellular cAMP in biochemical and cell signaling studies.

Catalog No. 50-002-35616 Supplier Medchemexpress LLC Supplier No. HY123185G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IBMX (3-isobutyl-1-methylxanthine) is a broad-spectrum phosphodiesterase (PDE) inhibitor used in research to raise intracellular cyclic AMP and probe signaling pathways. It is provided as a solid reagent for biochemical and cell-based assays.

  • Broad-spectrum phosphodiesterase inhibitor.
  • Inhibitory activity: PDE3 IC50 6.5 μM, PDE4 IC50 26.3 μM, PDE5 IC50 31.7 μM.
  • Molecular formula C10H14N4O2; molecular weight 222.24 g/mol.
  • Supplied as a powdered reagent suitable for research use.
  • Storage: powder -20°C for long-term storage, 4°C for shorter storage.
  • Reported purity approximately 99.97% by HPLC.
  • Used to elevate intracellular cAMP in biochemical and cell signaling studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.