Learn More
Medchemexpress LLC 3-isobutyl-1-methylxanthine | 28822-58-4 | MFCD00005584 | 222.24 g/mol | C10H14N4O2 | 5MG
SDP
Catalog No. 5000238260
Encompass_Preferred
IBMX (3-isobutyl-1-methylxanthine) is a broad-spectrum phosphodiesterase (PDE) inhibitor used as a research reagent and analytical standard. It raises intracellular cyclic AMP and cyclic GMP by inhibiting multiple PDE isoforms and is commonly used in biochemical and cell-based assays. Reported IC50 values reflect activity against several PDE families.
Catalog No. 50-002-38260 Supplier Medchemexpress LLC Supplier No. HY123185MG
IBMX (3-isobutyl-1-methylxanthine) is a broad-spectrum phosphodiesterase (PDE) inhibitor used as a research reagent and analytical standard. It raises intracellular cyclic AMP and cyclic GMP by inhibiting multiple PDE isoforms and is commonly used in biochemical and cell-based assays. Reported IC50 values reflect activity against several PDE families.