Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals 3-Isobutyl-1-methylxanthine, 99+%

Catalog No. AC228420010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC228420010 1 g Glass bottle
AC228420050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC228420010 Supplier Thermo Scientific Chemicals Supplier No. 228420010
  • $305.00
    $246.65 / Each of 1

    Save $58.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 28822-58-4 | C10H14N4O2 | 222.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28822-58-4
Molecular Formula C10H14N4O2
Molecular Weight (g/mol) 222.25
MDL Number MFCD00005584
InChI Key APIXJSLKIYYUKG-UHFFFAOYSA-N
Synonym 3-isobutyl-1-methylxanthine, ibmx, isobutylmethylxanthine, 1-methyl-3-isobutylxanthine, methylisobutylxanthine, 3-isobutyl-1-methyl-1h-purine-2,6 3h,7h-dione, xanthine, 3-isobutyl-1-methyl, 3-isobutyl-1-methyxanthine, 1h-purine-2,6-dione, 3,7-dihydro-1-methyl-3-2-methylpropyl, methyl-isobutylxanthine
PubChem CID 3758
ChEBI CHEBI:43253
SMILES CC(C)CN1C2=C(NC=N2)C(=O)N(C)C1=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 200.0°C to 203.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 1 g
Beilstein 26, IV, 2350
Solubility Information Solubility in water: almost insoluble. Other solubilities: soluble in ethanol,2-propanol
IUPAC Name 1-methyl-3-(2-methylpropyl)-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Formula Weight 222.25
Percent Purity 99+%
Physical Form Fluffy Crystalline Powder
Chemical Name or Material 3-Isobutyl-1-methylxanthine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 249-259-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.