Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Isobutyl-1-Methylxanthine, MP Biomedicals™

Catalog No. mp21952621 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21952621 100 mg
MP21952622 250 mg
MP219526201 1 g
MP219526205 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP21952621 Supplier MP Biomedicals, Inc Supplier No. 02195262.1
Add to Cart
Add to Cart

Known as IBMX. 3-Isobutyl-1-Methylxanthine, MP Biomedicals is a potent, non-specific inhibitor of cAMP and cGMP phosphodiesterases, significantly more effective than theophylline. This inhibition activates PKA, leading to decreased proliferation, increase

A non-specific inhibitor of adenosine 3',5'-cyclic monophosphate phosphodiesterase (cAMP PDE)4, significantly more effective than theophylline. Also inhibits cGMP phosphodiesterases. IBMX inhibits cyclic nucleotide PDE with subsequent inhibition of cyclic nucleotide hydrolysis, resulting in accumulation of cyclic AMP and guanosine 3',5'-cyclic monophosphate.

  • Inhibits phenylephrine-induced release of 5-hydroxytryptamine from neuroendocrine epithelial cells of the airway mucosa (IC50: 1.3μM)
  • Serves as an adenosine receptor antagonist
  • Inhibits ion channels in the neuromuscular junction, GH3 cells, and vascular smooth muscle cells
  • Induces calcium release from intracellular stores in sensory neurons
  • Inhibits the growth of carcinoma cells both in vivo and in vitro in mice
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28822-58-4
Molecular Formula C10H14N4O2
Molecular Weight (g/mol) 222.25
MDL Number MFCD00005584
InChI Key APIXJSLKIYYUKG-UHFFFAOYSA-N
Synonym 3-isobutyl-1-methylxanthine, ibmx, isobutylmethylxanthine, 1-methyl-3-isobutylxanthine, methylisobutylxanthine, 3-isobutyl-1-methyl-1h-purine-2,6 3h,7h-dione, xanthine, 3-isobutyl-1-methyl, 3-isobutyl-1-methyxanthine, 1h-purine-2,6-dione, 3,7-dihydro-1-methyl-3-2-methylpropyl, methyl-isobutylxanthine
PubChem CID 3758
ChEBI CHEBI:43253
IUPAC Name 1-methyl-3-(2-methylpropyl)-2,3,6,7-tetrahydro-1H-purine-2,6-dione
SMILES CC(C)CN1C2=C(NC=N2)C(=O)N(C)C1=O

Specifications

Melting Point 195°C to 202°C
Quantity 100 mg
Formula Weight 222.25
Physical Form Powder
Recommended Storage Store at 0°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.