Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Isocyanato-2-methyl-6-(trifluoromethyl)pyridine, Thermo Scientific™

Catalog No. MAY00326DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAY00326DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAY00326DA Supplier Thermo Scientific Chemicals Supplier No. MAY00326DA
Add to Cart

Chemical Identifiers

CAS 874832-10-7
Molecular Formula C8H5F3N2O
Molecular Weight (g/mol) 202.136
InChI Key BRXUOLWIERBAPA-UHFFFAOYSA-N
Synonym 3-isocyanato-2-methyl-6-trifluoromethyl pyridine, pyridine, 3-isocyanato-2-methyl-6-trifluoromethyl, 2-methyl-6-trifluoromethyl pyridin-3-isocyanate
PubChem CID 45594323
IUPAC Name 3-isocyanato-2-methyl-6-(trifluoromethyl)pyridine
SMILES CC1=C(C=CC(=N1)C(F)(F)F)N=C=O

Specifications

Quantity 1 g
Formula Weight 202.13
Chemical Name or Material 3-Isocyanato-2-methyl-6-(trifluoromethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.