Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Isocyanatopropyl)triethoxysilane, 95%

Catalog No. p-3593194
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC391900250 25 g Glass bottle
AC391901000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC391900250 Supplier Thermo Scientific Chemicals Supplier No. 391900250
Only null left
Add to Cart

CAS: 24801-88-5 | C10H21NO4Si | 247.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24801-88-5
Molecular Formula C10H21NO4Si
Molecular Weight (g/mol) 247.37
MDL Number MFCD00051459
InChI Key FRGPKMWIYVTFIQ-UHFFFAOYSA-N
Synonym 3-isocyanatopropyltriethoxysilane, triethoxy 3-isocyanatopropyl silane, 3-isocyanatopropyl triethoxysilane, isocyanatopropyltriethoxysilane, 3-triethoxysilyl propyl isocyanate, silane, triethoxy 3-isocyanatopropyl, unii-9br6002p6e, isocyanic acid, 3-triethoxysilyl propylester, isocyanic acid, 3-triethoxysilyl propyl ester, icptes
PubChem CID 90613
IUPAC Name triethoxy(3-isocyanatopropyl)silane
SMILES CCO[Si](CCCN=C=O)(OCC)OCC

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 130.0°C (20.0 mmHg)
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4190 to 1.4220 (20°C, 589nm)
Linear Formula (CH3CH2O)3Si(CH2)3NCO
Quantity 25 g
Specific Gravity 0.99
Solubility Information Solubility in water: reacts
Formula Weight 247.37
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material (3-Isocyanatopropyl)triethoxysilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
Fatal if inhaled.
Harmful if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
EINECSNumber 246-467-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.