Learn More
Medchemexpress LLC 3-Isoquinolinamine, N-[2-[(3S,4R)-3-fluoro-4-methoxy-1-piperidinyl]-4-pyrimidinyl]-5-(1-met | 2660250-10-0 | 99.4% | C28H37FN6O3S | 50 MG
SDP
Catalog No. 5000355347
Encompass_Preferred
BLU-945 is a potent, highly selective, reversible, and orally active epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor (TKI). It effectively inhibits EGFR with L858R and/or exon 19 deletion mutation, T790M mutation, and C797S mutation. This product is used for the research of lung cancer, including non-small cell lung cancer (NSCLC).
  • Potent and highly selective EGFR tyrosine kinase inhibitor
  • Reversible and orally active
  • Inhibits EGFR with L858R and/or exon 19 deletion mutation, T790M mutation, and C797S mutation
  • Used for research of lung cancer, including non-small cell lung cancer
  • Shows inhibitory activity against EGFRm/T790M double and EGFRm/T790M/C797S triple mutants
  • Demonstrates robust EGFR pathway inhibition and anti-tumor activity in triple-mutant Osimertinib-resistant models
Catalog No. 50-003-55347 Supplier Medchemexpress LLC Supplier No. HY14468050MG
BLU-945 is a potent, highly selective, reversible, and orally active epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor (TKI). It effectively inhibits EGFR with L858R and/or exon 19 deletion mutation, T790M mutation, and C797S mutation. This product is used for the research of lung cancer, including non-small cell lung cancer (NSCLC).
  • Potent and highly selective EGFR tyrosine kinase inhibitor
  • Reversible and orally active
  • Inhibits EGFR with L858R and/or exon 19 deletion mutation, T790M mutation, and C797S mutation
  • Used for research of lung cancer, including non-small cell lung cancer
  • Shows inhibitory activity against EGFRm/T790M double and EGFRm/T790M/C797S triple mutants
  • Demonstrates robust EGFR pathway inhibition and anti-tumor activity in triple-mutant Osimertinib-resistant models