Learn More
Medchemexpress LLC 3-Isoxazolecarboxamide, 5-(2-furanyl)-N-[2-(1H-pyrazol-1-yl)ethyl]- | 943820-93-7 | 99.7% | C13H12N4O3 | 50 MG
SDP
Catalog No. 5000354816
Encompass_Preferred
Wnt/β-catenin agonist 2 is a potent Wnt agonist that activates Wnt/β-catenin signaling. It can be used in the research of diseases related to signal transduction.
  • Activates Wnt/β-catenin signaling
  • Potent Wnt agonist
  • Used in research of diseases related to signal transduction
  • Activator for β-Catenin in SW480 cell line
  • Influences differentiation of ST2 cells into osteoblasts
  • White to off-white solid
Catalog No. 50-003-54816 Supplier Medchemexpress LLC Supplier No. HY14187350MG
Wnt/β-catenin agonist 2 is a potent Wnt agonist that activates Wnt/β-catenin signaling. It can be used in the research of diseases related to signal transduction.
  • Activates Wnt/β-catenin signaling
  • Potent Wnt agonist
  • Used in research of diseases related to signal transduction
  • Activator for β-Catenin in SW480 cell line
  • Influences differentiation of ST2 cells into osteoblasts
  • White to off-white solid