Learn More
Medchemexpress LLC 3-Isoxazolecarboxamide, 5-phenyl-N-[3-(2-thiazolyl)propyl]- | 2765218-56-0 | 99.1% | C16H15N3O2S | 25 MG
SDP
Catalog No. 5000357184
Encompass_Preferred
PY-60 is a robust and specific activator of YAP transcriptional activity that targets annexin A2 (ANXA2) with a Kd of 1.4 μM. It directly binds to ANXA2 and antagonizes its normal cellular function of repressing YAP activity, leading to increased levels of YAP-controlled transcripts in various cell lines. It can also activate a proproliferative, YAP-dependent transcriptional program in adult animals.
  • Activates YAP transcriptional activity
  • Targets annexin A2 (ANXA2) with a Kd of 1.4 μM
  • Directly binds to ANXA2 and antagonizes its normal function
  • Increases levels of YAP-controlled transcripts (ANRKD1, CYR61, and CTGF)
  • Promotes nuclear localization of YAP
  • Can remodel the epidermis through proliferation in adult animals
Catalog No. 50-003-57184 Supplier Medchemexpress LLC Supplier No. 25MGHY141644
PY-60 is a robust and specific activator of YAP transcriptional activity that targets annexin A2 (ANXA2) with a Kd of 1.4 μM. It directly binds to ANXA2 and antagonizes its normal cellular function of repressing YAP activity, leading to increased levels of YAP-controlled transcripts in various cell lines. It can also activate a proproliferative, YAP-dependent transcriptional program in adult animals.
  • Activates YAP transcriptional activity
  • Targets annexin A2 (ANXA2) with a Kd of 1.4 μM
  • Directly binds to ANXA2 and antagonizes its normal function
  • Increases levels of YAP-controlled transcripts (ANRKD1, CYR61, and CTGF)
  • Promotes nuclear localization of YAP
  • Can remodel the epidermis through proliferation in adult animals