Learn More
AdipoGen 3-ITCB-BPO-pyridinium bromide
SDP
Catalog No. 502560653
Encompass_Preferred
Chemical. CAS 155863-01-7. Formula C25H20BrN3O2S. MW 506.41. Synthetic. Class of highly photostable, water soluble fluorescent labels with large Stokes shift, high QY in aqueous media and pH tolerance. Relatively low extinction coefficient Epsilon∼ >20,000 in EtOH.
Catalog No. 50-256-0653 Supplier AdipoGen Supplier No. CDXI0034M025
Chemical. CAS 155863-01-7. Formula C25H20BrN3O2S. MW 506.41. Synthetic. Class of highly photostable, water soluble fluorescent labels with large Stokes shift, high QY in aqueous media and pH tolerance. Relatively low extinction coefficient Epsilon∼ >20,000 in EtOH.