Learn More
AdipoGen 3-ITCB-MPO-pyridinium bromide
SDP
Catalog No. 502560664
Encompass_Preferred
Chemical. CAS 155862-89-8. Formula C23H18BrN3O2S. MW 480.38. Synthetic. New class of highly photostable, water soluble fluorescent labels with large Stokes shift, high QY in aqueous media and pH tolerance.
Catalog No. 50-256-0664 Supplier AdipoGen Supplier No. CDXI0022M250
Chemical. CAS 155862-89-8. Formula C23H18BrN3O2S. MW 480.38. Synthetic. New class of highly photostable, water soluble fluorescent labels with large Stokes shift, high QY in aqueous media and pH tolerance.