Learn More
AdipoGen 3-ITCEt-BPO-pyridinium bromide
SDP
Catalog No. 502560648
Encompass_Preferred
Chemical. CAS 155863-02-8. Formula C20H18BrN3O2S. MW 444.34. Synthetic. Class of highly photostable, water soluble fluorescent labels with large Stokes shift, high QY in aqueous media and pH tolerance. Relatively low extinction coefficient Epsilon∼ >20,000 in EtOH.
Catalog No. 50-256-0648 Supplier AdipoGen Supplier No. CDXI0035M250
Chemical. CAS 155863-02-8. Formula C20H18BrN3O2S. MW 444.34. Synthetic. Class of highly photostable, water soluble fluorescent labels with large Stokes shift, high QY in aqueous media and pH tolerance. Relatively low extinction coefficient Epsilon∼ >20,000 in EtOH.