Learn More
Cayman Chemical 3-keto SphngnInd18 0h 10mg
SDP
Catalog No. 502936029
Encompass_Preferred
An intermediate in the biosynthesis of C18 ceramide; a lyso-sphingolipid formed by the condensation of L-serine and palmitoyl-CoA by SPT; vitamin K deficiency decreases production by SPT
Catalog No. 50-293-6029 Supplier Cayman Chemical Supplier No. 2438010 MG
An intermediate in the biosynthesis of C18 ceramide; a lyso-sphingolipid formed by the condensation of L-serine and palmitoyl-CoA by SPT; vitamin K deficiency decreases production by SPT