Learn More
Medchemexpress LLC 3-ketoadipic acid | 689-31-6 | MFCD00010003 | 95.0% | 160.12 g/mol | C6H8O5 | 1mg
SDP
Catalog No. 5000270489
Encompass_Preferred
3-Ketoadipic acid is a dicarboxylic acid containing a keto group that can be isolated from Brazilian propolis (Et-Bra) Brazilian propolis exhibits antibacterial antifungal and antiparasitic activities[1]
Catalog No. 50-002-70489 Supplier Medchemexpress LLC Supplier No. HYW7184181MG
3-Ketoadipic acid is a dicarboxylic acid containing a keto group that can be isolated from Brazilian propolis (Et-Bra) Brazilian propolis exhibits antibacterial antifungal and antiparasitic activities[1]