Learn More
Abcam 3-Ketodihydrosphingosine hydrochloride, Ceramide biosynthesis intermediate, 10MG
Catalog No. 502706030
Encompass_Preferred
MW 335.96 g/mol, Purity >98%. Necessary intermediate in ceramide (ab141745, ab141743) biosynthesis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6030 Supplier Abcam Supplier No. AB14410710MG
MW 335.96 g/mol, Purity >98%. Necessary intermediate in ceramide (ab141745, ab141743) biosynthesis.
The product is subject to the following: Abcam Restricted Use Statement