Learn More
Apexbio Technology LLC 3-KETOLITHOCHOLIC-ACID-1MG
SDP
Catalog No. 5000567554
Encompass
3-Ketolithocholic acid (CAS No 1553-56-6) is a bile acid derivative commonly studied in biomedical research for its role in metabolic and liver disorders It originates from the oxidation of lithocholic acid and is known to interact with nuclear receptors and signaling pathways involved in lipid metabolism such as the farnesoid X receptor (FXR) Its in vitro activity is typically observed in the nanomolar to low micromolar range indicating potential efficacy in modulating these pathways Common applications include studies in cell models and animal research particularly focusing on liver health and cholesterol regulation It is also utilized in drug screening to assess the therapeutic effects on metabolic pathways Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-67554 Supplier Apexbio Technology LLC Supplier No. C86391
Add to Cart
Add to Cart
3-Ketolithocholic acid (CAS No 1553-56-6) is a bile acid derivative commonly studied in biomedical research for its role in metabolic and liver disorders It originates from the oxidation of lithocholic acid and is known to interact with nuclear receptors and signaling pathways involved in lipid metabolism such as the farnesoid X receptor (FXR) Its in vitro activity is typically observed in the nanomolar to low micromolar range indicating potential efficacy in modulating these pathways Common applications include studies in cell models and animal research particularly focusing on liver health and cholesterol regulation It is also utilized in drug screening to assess the therapeutic effects on metabolic pathways Experimental concentrations typically depend on the specific research design