Learn More
Chemglass Life Sciences Beaker, Low Form, 3000mL, Griffin, Double Scale, Graduated
SDP
Catalog No. NC0408428
Encompass_Preferred
Beaker, Low Form, 3000mL, Griffin, Double Scale, Graduated, Beaker, with spout, manufactured with uniform wall thickness, offers optimum balance between thermal shock resistance and mechanical strength. For convenience, the 250 through 4000mL beakers have a double graduated metric scale to indicate approximate content. All sizes have an extra large marking spot. The 10mL size is not graduated.
This item is not returnable. View return policy
Catalog No. NC0408428 Supplier Chemglass Life Sciences Supplier No. CG80483L
Beaker, Low Form, 3000mL, Griffin, Double Scale, Graduated, Beaker, with spout, manufactured with uniform wall thickness, offers optimum balance between thermal shock resistance and mechanical strength. For convenience, the 250 through 4000mL beakers have a double graduated metric scale to indicate approximate content. All sizes have an extra large marking spot. The 10mL size is not graduated.