Learn More
International Enviroguard™ 3-Layer Latex Free Disposable Masks
Catalog No. 19472482 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Construction:
3-Ply
Catalog No. Construction
19-472-482 3-Ply
1 options
Catalog No. 19-472-482 Supplier International Enviroguard™ Supplier No. 2804
Add to Cart
Add to Cart
3-Layer Disposable Masks are latex-free, breathable and provide 99% bacterial filtration efficiency and a pliable nose clip for a secure fit.
These latex-free pleated masks are made with a 3-ply material that provides high bacterial filtration efficiency (BFE) with maximum comfort. With up to 99% bacterial filtration, you can work with peace of mind all shift long. Better yet, these masks are designed with soft-fit features for added comfort and excellent breathability. Simple earloops offer easy donning without messy ties. Loops are made of a comfortable spandex/polyester blend. Ultrasonically bonded edges and secure elastic on both sides ensure this mask holds up as you put in a hard day's work. A pliable nose clip allows for an adjustable, more secure fit to your face for enhanced protection.
  • Outstanding bacterial filtration (99%)
  • Pliable nose clip that adjusts for a secure fit
  • Secure elastic on both sides
  • Ultrasonically bonded edges provide improved product strength and integrity
  • Latex/Fiberglass free
  • General-use masks are not intended for use by Healthcare Professionals
Specifications
Product Type General Purpose Face Mask
Color Light Blue
Fastening Type Earloops
Material Polypropylene
Size Standard
Sterility Non-sterile
Style Pleated
Construction 3-Ply
Disposable Single-use
Filter Efficiency 99% (BFE)
Latex Free Yes
Model 2804