Learn More
Edmund Optics 3" Length Box (10 Bags), Cotton-Tipped Swabs, 1000/cs
Catalog No. NC1701979
Encompass
These lint-free swabs, with 3" or 6" long wooden stems, are ideal for optical clean-up applications and useful for a variety of other applications. Non-sterile.
Catalog No. NC1701979 Supplier Edmund Optics Supplier No. 56927
These lint-free swabs, with 3" or 6" long wooden stems, are ideal for optical clean-up applications and useful for a variety of other applications. Non-sterile.