Learn More
Chemglass Life Sciences Tripod Stand for 1L, 2L, & 3L Unjacketed Bio Reactor Vessels
SDP
Catalog No. 501294468
Encompass
Tripod Stand for 2-3L Unjacketed Bio Reactor Vessels. Stand to hold unjacketed bio reaction vessels. Top, two-piece ring is made of anodized aluminum while the lower ring and legs are electropolished stainless steel. Stands come complete with silicone o-ring and six, knurled nuts to attach the headplate to the stand.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
This item is not returnable. View return policy
Catalog No. 50-129-4468 Supplier Chemglass Life Sciences Supplier No. CLS1380STD3
Tripod Stand for 2-3L Unjacketed Bio Reactor Vessels. Stand to hold unjacketed bio reaction vessels. Top, two-piece ring is made of anodized aluminum while the lower ring and legs are electropolished stainless steel. Stands come complete with silicone o-ring and six, knurled nuts to attach the headplate to the stand.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.