Learn More
Ace Glass, Inc. Beaker, 3L, jacketed, Ace-Safe hose connections for 1/2intubing, 225mm inside height, 133mm inside diameter
SDP
Catalog No. NC9262469
Encompass_Preferred
3L jacketed beaker with one upper and one lower Ace-Safe hose connection on opposite sides, for 1/2 in. tubing. Dimensions: 225mm I.H. x 133mm I.D. Does not have a pouring spout.
This item is not returnable. View return policy
Catalog No. NC9262469 Supplier Ace Glass, Inc. Supplier No. 5340125
3L jacketed beaker with one upper and one lower Ace-Safe hose connection on opposite sides, for 1/2 in. tubing. Dimensions: 225mm I.H. x 133mm I.D. Does not have a pouring spout.