Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Maleimidobenzoic Acid N-Hydroxysuccinimide Ester

Catalog No. M278625MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
12 x 6 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M278625MG 100 mg
M2786100MG 12 x 6 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M278625MG Supplier Sigma Aldrich Supplier No. M278625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Maleimidobenzoic Acid N-Hydroxysuccinimide Ester

Specifications

CAS 58626-38-3
Melting Point 175°C to 177°C
Molecular Formula C15H10N2O6
Linear Formula C15H10N2O6
MDL Number MFCD00005514
Quantity 100 mg
Synonym MBS
Molecular Weight (g/mol) 314.25 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.