Learn More
Apexbio Technology LLC 3-Maleimidopropionic-acid
SDP
Catalog No. 5000569407
Encompass_Preferred
3-Maleimidopropionic acid (CAS No 7423-55-4) is a synthetic compound commonly used in the field of biomedical research particularly for its role in modifying and cross-linking proteins and peptides Originating from the modification of maleimide chemistry it acts primarily by targeting thiol groups forming stable thioether bonds This specificity is invaluable for studying protein-protein interactions and drug binding dynamics Its activity is generally observed in the nanomolar to low micromolar range making it suitable for precise biochemical assays Typically it finds applications in cell culture studies animal models and high-throughput drug screening where it is used to investigate cellular pathways and potential therapeutic targets Experimental concentrations typically depend on the specific research design ensuring flexibility and adaptability in various experimental setups
Catalog No. 50-005-69407 Supplier Apexbio Technology LLC Supplier No. C7034500
3-Maleimidopropionic acid (CAS No 7423-55-4) is a synthetic compound commonly used in the field of biomedical research particularly for its role in modifying and cross-linking proteins and peptides Originating from the modification of maleimide chemistry it acts primarily by targeting thiol groups forming stable thioether bonds This specificity is invaluable for studying protein-protein interactions and drug binding dynamics Its activity is generally observed in the nanomolar to low micromolar range making it suitable for precise biochemical assays Typically it finds applications in cell culture studies animal models and high-throughput drug screening where it is used to investigate cellular pathways and potential therapeutic targets Experimental concentrations typically depend on the specific research design ensuring flexibility and adaptability in various experimental setups