Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-maleimidopropionic acid N-hydroxysuccinimide ester | 55750-62-4 | MFCD00043141 | 99.9% | 266.21 g/mol | C11H10N2O6 | 50 G
SDP

Catalog No. 5000202308
Encompass_Preferred

BMPS is a non-cleavable antibody-drug conjugate (ADC) linker used to connect cytotoxic payloads to antibodies during ADC synthesis. It is supplied as a solid for research use only and is characterized by a maleimide-based reactive group for selective conjugation to thiol residues.

  • Non-cleavable maleimide-based linker for stable ADC conjugation.
  • Chemical name: 3-maleimidopropionic acid N-hydroxysuccinimide ester.
  • CAS number: 55750-62-4.
  • Molecular weight: 266.21 g/mol; formula C11H10N2O6.
  • Reported purity 99.93% (listed as 99.9% rounded).
  • Available as a solid in a 50 G pack.
  • Intended for research use only; handle according to laboratory safety guidance.

Catalog No. 50-002-02308 Supplier Medchemexpress LLC Supplier No. HY4214650G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

BMPS is a non-cleavable antibody-drug conjugate (ADC) linker used to connect cytotoxic payloads to antibodies during ADC synthesis. It is supplied as a solid for research use only and is characterized by a maleimide-based reactive group for selective conjugation to thiol residues.

  • Non-cleavable maleimide-based linker for stable ADC conjugation.
  • Chemical name: 3-maleimidopropionic acid N-hydroxysuccinimide ester.
  • CAS number: 55750-62-4.
  • Molecular weight: 266.21 g/mol; formula C11H10N2O6.
  • Reported purity 99.93% (listed as 99.9% rounded).
  • Available as a solid in a 50 G pack.
  • Intended for research use only; handle according to laboratory safety guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.