Learn More
Medchemexpress LLC 3-MeOARh-NTR | 596.61 | 5 MG
SDP
Catalog No. 5000360073
Encompass_Preferred
3-MeOARh-NTR is an activatable imaging probe with high selectivity and good stability, designed for nitroreductase (NTR) detection. It offers a high signal-to-noise ratio, making it an efficient molecular tool for endogenous NTR detection and useful for evaluating kidney hypoxia.
  • Produces a strong fluorescence signal in living HeLa cells with decreasing oxygen contents
  • Produces fluorescence imaging of kidney tissues from mice with excitation = 488 nm and emission = 510-590 nm
Catalog No. 50-003-60073 Supplier Medchemexpress LLC Supplier No. HY1498365MG
3-MeOARh-NTR is an activatable imaging probe with high selectivity and good stability, designed for nitroreductase (NTR) detection. It offers a high signal-to-noise ratio, making it an efficient molecular tool for endogenous NTR detection and useful for evaluating kidney hypoxia.
  • Produces a strong fluorescence signal in living HeLa cells with decreasing oxygen contents
  • Produces fluorescence imaging of kidney tissues from mice with excitation = 488 nm and emission = 510-590 nm